missing translation for 'onlineSavingsMsg'
Learn More

Puerarin, TRC

Product Code. 30005119
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005119 10 mg 10mg
30136797 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005119 Supplier Toronto Research Chemicals Supplier No. TRCP84013010MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3681-99-0
Molecular Formula C21 H20 O9
Molecular Weight (g/mol) 416.38
Synonym 4H-1-Benzopyran-4-one, 8-β-D-glucopyranosyl-7-hydroxy-3-(4-hydroxyphenyl)-, Puerarin (6CI,7CI,8CI), 8-β-D-Glucopyranosyl-7-hydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Daidzein 8-C-glucoside, Kakonein, NPI 031G
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)ccc3C(=O)C(=COc23)c4ccc(O)cc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H20O9/c22-7-14-17(26)18(27)19(28)21(30-14)15-13(24)6-5-11-16(25)12(8-29-20(11)15)9-1-3-10(23)4-2-9/h1-6,8,14,17-19,21-24,26-28H,7H2/t14-,17-,18+,19-,21+/m1/s1
Formula Weight 416.1107
Percent Purity >95%
Chemical Name or Material Puerarin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.