Learn More
Pyridine-d5 (Reagent grade), CDN
Product Code. 30327981
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327981 5 g 5g
30328208 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327981 Supplier CDN Supplier No. CDND00855G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyridine D5
CAS 7291-22-7
Molecular Formula C5 2H5 N
Quantity 5 g
Synonym Pyridine-2,3,4,5,6-d5, Pyridine-d5 (6 CI,7 CI,8 CI,9 CI), Pentadeuteropyridine, Perdeuteriopyridine
InChI Formula InChI=1 S/C5H5N/c1-2-4-6-5-3-1/h1-5 H/i1D,2 D,3 D,4 D,5 D
SMILES [2 H]c1nc([2 H])c([2 H])c([2 H])c1[2 H]
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
Molecular Weight (g/mol) 84.13
Formula Weight 84.0736 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity