missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium Chlorochromate, TRC

Product Code. 30411609
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411609 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411609 Supplier Toronto Research Chemicals Supplier No. TRCP991820100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H5N.ClCrO3.H
Molecular Weight (g/mol) 215.56
Synonym Chlorotrioxochromate(1-) Hydrogen, compd. with Pyridine (1:1), Chlorotrioxochromate(1-)pyridine, Trioxochlorochromate(VI)Pyridine, Chlorotrioxochromate(1-) Hydrogen, compd. With Pyridine (1:1), Pyridine, (T-4)-chlorotrioxochromate(1-), Corey Reagent, Corey's reagent, Corey's Reagent (oxidation reagent), PCC, PCC (oxidizing agent), Pyridinium Chlorochromate, Reagents, Corey's, Urine Luck
IUPAC Name chloro-hydroxy-dioxochromium;pyridine
SMILES O[Cr](=O)(=O)Cl.c1ccncc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H5N.ClH.Cr.3O/c1-2-4-6-5-3-1;;;;;/h1-5H;1H;;;;
Formula Weight 214.9441
Chemical Name or Material Pyridinium Chlorochromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.