missing translation for 'onlineSavingsMsg'
Learn More

Pyriofenone, TRC

Product Code. 30133248
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133248 5 mg 5mg
17942245 10 mg 10mg
17922605 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133248 Supplier Toronto Research Chemicals Supplier No. TRCP9973705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688046-61-9
Molecular Formula C18H20ClNO5
Molecular Weight (g/mol) 365.81
Synonym (5-Chloro-2-methoxy-4-methyl-3-pyridinyl)(2,3,4-trimethoxy-6-methylphenyl)methanone, 3-(2,3,4-Trimethoxy-6-methylbenzoyl)-5-chloro-2-methoxy-4-methylpyridine, Chlazafenone, Clazafenone, IKF 309, Property Flowable, Pyriofenone
IUPAC Name (5-chloro-2-methoxy-4-methylpyridin-3-yl)-(2,3,4-trimethoxy-6-methylphenyl)methanone
SMILES COc1cc(C)c(C(=O)c2c(C)c(Cl)cnc2OC)c(OC)c1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H20ClNO5/c1-9-7-12(22-3)16(23-4)17(24-5)13(9)15(21)14-10(2)11(19)8-20-18(14)25-6/h7-8H,1-6H3
Formula Weight 365.103
Percent Purity >95%
Chemical Name or Material Pyriofenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.