missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-2-carbonitrile, 97%

Product Code. 11388726
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388726 1 g 1g
11398726 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388726 Supplier Thermo Scientific Alfa Aesar Supplier No. L19427.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1436-43-7 | C10H6N2 | 154.17 g/mol

Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-43-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00134341
InChI Key WDXARTMCIRVMAE-UHFFFAOYSA-N
Synonym 2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
PubChem CID 74031
IUPAC Name quinoline-2-carbonitrile
SMILES N#CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 93°C to 96°C
Boiling Point 160°C (23 mmHg)
Odor Odorless
Quantity 1 g
Beilstein 115213
Solubility Information Insoluble in water.
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material Quinoline-2-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.