missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab3D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3337-0.025mg
This item is not returnable.
View return policy
Description
Rab3D Polyclonal specifically detects Rab3D in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Rab3D | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RAB3D | |
| Synthetic peptide corresponding to residues M(1) A S A S E P P A S P R D A A D Q N(18) of rat RAB3D. | |
| 0.025 mg | |
| Membrane Trafficking and Chaperones, Signal Transduction | |
| 9545 | |
| Mouse | |
| Purified |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2.0 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| D2-2, GOVras-related protein Rab-3D, RAB16glioblastoma overexpressed, RAB3D, member RAS oncogene family, RAD3DRab3D upregulated with myeloid differentiation | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Detects recombinant mouse rab 3D. This does not detect rab 3A, 3B, or 3C. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction