missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB6B Antibody (6D12E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61825-0.025ml
This item is not returnable.
View return policy
Description
RAB6B Monoclonal specifically detects RAB6B in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RAB6B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| RAB6B, member RAS oncogene family, ras-related protein Rab-6B, small GTPase RAB6B, small GTP-binding protein | |
| Mouse | |
| 23.5 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 6D12E4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Rab6b | |
| Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 51560 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction