missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB6B Antibody (6D12E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 552.00€
Specifications
| Antigen | RAB6B |
|---|---|
| Clone | 6D12E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204272
|
Novus Biologicals
NBP2-61825 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679418
|
Novus Biologicals
NBP2-61825-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB6B Monoclonal specifically detects RAB6B in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RAB6B | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| RAB6B, member RAS oncogene family, ras-related protein Rab-6B, small GTPase RAB6B, small GTP-binding protein | |
| Rab6b | |
| IgG1 | |
| Protein G purified |
| 6D12E4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 51560 | |
| Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. Coli. | |
| Primary | |
| 23.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title