missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB9 Monoclonal Antibody (Mab9)
GREENER_CHOICE

Product Code. 11525762
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11525762 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11525762 Supplier Invitrogen™ Supplier No. MA3067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA3-067 recognizes prenylated and non-prenylated rab 9 from canine, human, monkey, rat, hamster and bovine tissues. This antibody does not cross-react with other rab family members. MA3-067 has been successfully used in Western blot, immunofluorescence, EM immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ~26 kDa protein representing rab 9 from K562 cell extract. The MA3-067 antigen is recombinant canine rab 9a.

Rab proteins are a family of Ras-like GTPases involved in intracellular compartment protein transport. Different members of the 40+ member rab family are responsible for docking and fusion of transport vesicles between different compartments within the cell. Rab 9 is required for trafficking mannose 6-phosphate receptor between the late endosome to trans-Golgi network (TGN). By facilitating receptor transport, rab 9 enables cells to efficiently recycle important cellular trafficking components. It is functionally necessary for rab 9, like other rab family members, to be prenylated by two 20-carbon geranylgeranyl groups at the C-terminus. Most prenylated rab 9 is membrane bound, however, 10-20% of the cellular pool of rab 9 is bound to GDP dissociation inhibitor-alpha (GDI-alpha) in the cytosol. GDI recycles prenylated, GDP bound rab 9 from their fusion targets back to their membranes of origin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB9
Applications Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Mab9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Rab9a
Gene Accession No. P24408, P51151, Q99P75
Gene Alias 2410064E05Rik; AI195561; rab9; rab9 GTP-binding protein; RAB9, member RAS oncogene family; Rab9a; rab-9A; RAB9A, member RAS oncogene family; RAB9-like protein; ras-related protein Rab-9A; Sid 99; Sid6061p; Sid99
Gene Symbols Rab9a
Host Species Mouse
Immunogen Recombinant canine rab 9a.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100767493, 403947, 511776, 84589, 9367
Target Species Bovine, Canine, Hamster, Human, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.