Learn More
NTRK2 (phospho Y706/Y707), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Description
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | NTRK2 |
| Applications | Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NTRK2. |
| Dilution | Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) |
| Gene | NTRK2 |
| Gene Alias | GP145-TrkB/TRKB |
| Gene Symbols | NTRK2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.