missing translation for 'onlineSavingsMsg'
Learn More

Rabies Virus Nucleoprotein Antibody (02) [FITC] - (CVS-11), Novus Biologicals™

Product Code. 30174687
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30174687 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30174687 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP333165F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rabies Virus Nucleoprotein Monoclonal antibody specifically detects Rabies Virus Nucleoprotein in Virus samples. It is validated for ELISA,Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus Nucleoprotein
Applications ELISA, Sandwich ELISA Capture
Classification Monoclonal
Clone 02
Conjugate FITC
Formulation PBS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabies Virus Nucleoprotein (Uniprot#: Q8JXF6; Met1-Ser450).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.