missing translation for 'onlineSavingsMsg'
Learn More

rac-2-Aminobutyric Acid-d3 Hydrochloride, TRC

Product Code. 30313727
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313727 25 mg 25mg
30322679 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30313727 Supplier Toronto Research Chemicals Supplier No. TRCA60293725MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-2-Aminobutyric Acid-d3 Hydrochloride
Molecular Formula C4H6D3NO2.HCl
Quantity 25 mg
Synonym (+/-)-2-Aminobutanoic Acid-d3 Hydrochloride, (+/-)-α-Aminobutyric Acid-d3 Hydrochloride, DL-α-2-Aminobutyric Acid-d3 Hydrochloride, DL-Butyrine-d3 Hydrochloride, DL-Ethylglycine-d3 Hydrochloride, Homoalanine Hydrochloride, NSC 3251-d3 Hydrochloride
InChI Formula InChI=1S/C4H9NO2.Cl/c1-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H,6,7);/i1D3;
SMILES NC(C(O)=O)CC([2H])([2H])[2H].Cl
IUPAC Name 2-aminobutanoic-4,4,4-d3 acid hydrochloric acid
Molecular Weight (g/mol) 142.6
Formula Weight 142.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.