missing translation for 'onlineSavingsMsg'
Learn More

rac N,O-Didesmethyl Venlafaxine, TRC

Product Code. 30138661
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138661 50 mg 50mg
17994935 10 mg 10mg
17914545 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138661 Supplier Toronto Research Chemicals Supplier No. TRCD44157550MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135308-74-6
Molecular Formula C15 H23 N O2
Molecular Weight (g/mol) 249.35
Synonym Phenol, 4-[1-(1-hydroxycyclohexyl)-2-(methylamino)ethyl]-, 4-[1-(1-Hydroxycyclohexyl)-2-(methylamino)ethyl]phenol, N,O-Didesmethylvenlafaxine, Venlafaxine-N,O-didesmethyl
IUPAC Name 4-[1-(1-hydroxycyclohexyl)-2-(methylamino)ethyl]phenol
SMILES CNCC(c1ccc(O)cc1)C2(O)CCCCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H23NO2/c1-16-11-14(12-5-7-13(17)8-6-12)15(18)9-3-2-4-10-15/h5-8,14,16-18H,2-4,9-11H2,1H3
Formula Weight 249.1729
Percent Purity >95%
Chemical Name or Material D,L N,O-Didesmethyl Venlafaxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.