Learn More
RAD1 Mouse, anti-Human, Clone: 1A11E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68637-1-IG-20UL
Description
The RAD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAD1. This antibody recognizes human antigen. The RAD1 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| RAD1 | |
| 1A11E2 | |
| HRAD1, RAD1, RAD1 homolog (S. pombe), REC1 | |
| 20 μL | |
| 5810 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunoprecipitation, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |