missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAP1 Monoclonal Antibody (4c8/1)
GREENER_CHOICE

Product Code. 11515572
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11515572 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11515572 Supplier Invitrogen™ Supplier No. MA191542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Human Rap1 was the first identified ortholog of the yeast telomeric protein scRap1, which shares 3 conserved sequence motifs in common. HRap1 is located at telomeres and affects telomere length by being recruited to telomeres by TRF2 instead of directly binding to the telomere like scRap1. The hRap1 cDNA codes for a protein of 399 amino acids with approximate molecular weight of 47 kDa. The hRap1 protein does not interact with TRF1. TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length(1). TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4c8/1
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TERF2IP
Gene Accession No. Q9NYB0
Gene Alias dopamine receptor interacting protein 5; dopamine receptor-interacting protein 5; DRIP5; hRap1; MNCb-0448; MNCb-0628; PP8000; RAP1; RAP1 homolog; repressor/activator protein 1 homolog; tel; telomeric repeat binding factor 2, interacting protein; telomeric repeat-binding factor 2-interacting protein 1; TERF2 interacting protein; TERF21P; TERF2-interacting telomeric protein 1; TERF2IP; TRF2-interacting telomeric protein 1; TRF2-interacting telomeric protein Rap1; TRF2-interacting telomeric RAP1 protein
Gene Symbols TERF2IP
Host Species Mouse
Immunogen Human Rap1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54386
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.