missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RDBP Recombinant Rabbit Monoclonal Antibody (K01_1U65)
Description
RDBP Recombinant Monoclonal Antibody for IHC (P), Western Blot
RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
Spécification
Spécification
| Antigen | RDBP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1U65 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | NELFE |
| Gene Accession No. | P18615, P19426 |
| Gene Alias | D17H6S45; D6S45; major histocompatibility complex gene RD; negative elongation factor complex member E; negative elongation factor complex member E, Rdbp; negative elongation factor E; negative elongation factor polypeptide E; NELF E; Nelfe; NELF-E; nuclear protein; Rd; RD RNA binding protein; RD RNA-binding protein; RDBP; RDP; RNA-binding protein RD |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?