missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REC8 Polyclonal Antibody

Description
The antibody detects endogenous levels of total REC8 protein.
This gene encodes a member of the kleisin family of SMC (structural maintenance of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigen | REC8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | REC8 |
| Gene Accession No. | O95072 |
| Gene Alias | cohesin Rec8p; cohesion rec8p; HR21spB; human homolog of rad21, S. pombe; kleisin-alpha; Mei8; meiosis specific sister chromatid cohesion protein; meiotic cohesion Rec8; meiotic recombination and sister chromatid cohesion phosphoprotein of the rad21p family; meiotic recombination protein REC8 homolog; meiotic recombination protein REC8-like 1; mrec; Rec8; rec8 {ECO:0000250; REC8 homolog; REC8 meiotic recombination protein; Rec8L1; REC8-like 1; Rec8p; recombination and sister chromatid cohesion protein homolog; UniProtKB:Q8C5S7} |
| Gene Symbols | REC8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?