missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REG4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16557061
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16557061 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16557061 Supplier Invitrogen™ Supplier No. PA589008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DIIMRPSCAP GWFYHKSNCY GYFRKLRNWS DAELECQSYG NGAHLASILS LKEASTIAEY ISGYQRSQPI WIGLHDPQKR QQWQWIDGAM YLYRSWSGKS MGGNKHCAEM SSNNNFLTWS SNECNKRQHF LCKYRP; Positive Samples: Mouse stomach, Rat stomach.

Reg IV is part of the regenerating gene family within the C-type lectin superfamily. This family is involved in liver, pancreatic, gastric and intestinal cell proliferation and differentiation. Reg IV is a 158-amino acid secretory protein implicated in cell regeneration and/or survival with a definite growth stimulating effect on pancreatic beta cells. It is highly expressed in colorectal, gastric, prostate and other types of cancer. Reg IV-positive tumor cells display different phenotypes including mucus-secreting, enterocyte-like, and undifferentiated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene REG4
Gene Accession No. Q68AX7, Q9D8G5
Gene Alias 2010002L15Rik; Gastrointestinal secretory protein; GISP; GISPREG-4; Reg IV; REG4; REG-4; regenerating family member 4; regenerating gene type IV; regenerating islet-derived family, member 4; regenerating islet-derived protein 4; Regenerating islet-derived protein IV; REG-IV; REG-like protein; RELP
Gene Symbols REG4
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445583, 67709
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.