missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RENALASE Mouse anti-Human, Clone: 3A12F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60128-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to RNLS.
Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.Specifications
| RENALASE | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VYX0 | |
| RNLS | |
| RENALASE Fusion Protein Ag13061 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3A12F5 | |
| Unconjugated | |
| RNLS | |
| C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS | |
| Mouse | |
| Protein G | |
| RUO | |
| 55328 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction