missing translation for 'onlineSavingsMsg'
Learn More

RENALASE Mouse anti-Human, Clone: 3A12F5, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen RENALASE
Clone 3A12F5
Concentration 0.6 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16867095
View Documents
Proteintech
60128-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16857095
View Documents
Proteintech
60128-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to RNLS.

Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.
Specifications

Specifications

RENALASE
0.6 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS
RNLS
IgG1
Protein G
Antibody
3A12F5
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q5VYX0
55328
RENALASE Fusion Protein Ag13061
Primary
-20°C
RNLS
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.