missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REPIN1 Recombinant Rabbit Monoclonal Antibody (2X2E9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557992

 View more versions of this product

Product Code. 30284131

  • 457.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SCDDCGRSFR LERFLRAHQR QHTGERPFTC AECGKNFGKK THLVAHSRVH SGERPFACEE CGRRFSQGSH LAAHRRDHAP DRPFVCPDCG KAFRHKPYLA AHRRIHTGEK PYVCPDCGKA FSQKSNLVSH RRIHTGERPY ACPDCDRSFS QKSNLITHRK SHIRDGAFCC AICGQTFDDE ERLLAHQKKH DV.

Replication initiator 1 (REPIN1) is a gene encoding a zinc-finger protein that plays a critical role in various metabolic processes. Located on chromosome 7 in humans, REPIN1 is prominently expressed in liver and adipose tissues. It is associated with the regulation of body weight and triglyceride levels, as evidenced by its locus within a quantitative trait locus (QTL) related to these metabolic traits. The REPIN1 protein is involved in enhancing DNA bending, which is crucial for replication initiation. Studies have shown that deleting REPIN1 in the liver can lead to improved insulin sensitivity and altered lipid metabolism, with secondary effects on adipose tissue. Additionally, REPIN1 is implicated in iron metabolism and apoptosis regulation, further underscoring its multifaceted role in cellular processes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

REPIN1
Recombinant Monoclonal
1.01 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q9BWE0
REPIN1
Recombinant fusion protein containing a sequence corresponding to amino acids 376-567 of human REPIN1 (NP_037532.2).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
2X2E9
Unconjugated
REPIN1
60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; REPIN1; replication initiation region protein (60kD); replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464
Rabbit
Protein A
RUO
29803
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.