Learn More
Invitrogen™ REPIN1 Recombinant Rabbit Monoclonal Antibody (2X2E9)

Rabbit Recombinant Monoclonal Antibody
176.00€ - 436.00€
Specifications
| Antigen | REPIN1 |
|---|---|
| Clone | 2X2E9 |
| Concentration | 1.01 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284436
|
Invitrogen™
MA557991 |
20 μL |
176.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284131
![]() |
Invitrogen™
MA557992 |
100 μL |
436.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: SCDDCGRSFR LERFLRAHQR QHTGERPFTC AECGKNFGKK THLVAHSRVH SGERPFACEE CGRRFSQGSH LAAHRRDHAP DRPFVCPDCG KAFRHKPYLA AHRRIHTGEK PYVCPDCGKA FSQKSNLVSH RRIHTGERPY ACPDCDRSFS QKSNLITHRK SHIRDGAFCC AICGQTFDDE ERLLAHQKKH DV.
Specifications
| REPIN1 | |
| 1.01 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9BWE0 | |
| 29803 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 376-567 of human REPIN1 (NP_037532.2). | |
| Antibody |
| 2X2E9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| REPIN1 | |
| 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; REPIN1; replication initiation region protein (60kD); replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464 | |
| REPIN1 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.