missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13503-1-AP-20UL
This item is not returnable.
View return policy
Description
Seems to have no detectable effect on transcription elongation in vitro. Tamura K., Biochem. Biophys. Res. Commun. 309:189-195(2003). Hirosawa M., DNA Res. 6:329-336(1999). The MGC Project Team; Genome Res. 14:2121-2127(2004).Specifications
| REX1 | |
| Polyclonal | |
| Unconjugated | |
| REXO1 | |
| EloA BP1, ELOABP1, Elongin A binding protein 1, KIAA1138, REX1, REXO1, RNA exonuclease 1 homolog, TCEB3BP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 314630, 57455, 66932 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TT28, Q8N1G1 | |
| Rexo1 | |
| REX1 Fusion Protein Ag4334 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction