missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | REX1 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16881744
|
Proteintech
13503-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16871744
|
Proteintech
13503-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Seems to have no detectable effect on transcription elongation in vitro. Tamura K., Biochem. Biophys. Res. Commun. 309:189-195(2003). Hirosawa M., DNA Res. 6:329-336(1999). The MGC Project Team; Genome Res. 14:2121-2127(2004).Specifications
| REX1 | |
| Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q7TT28, Q8N1G1 | |
| 314630, 57455, 66932 | |
| REX1 Fusion Protein Ag4334 | |
| Primary | |
| -20°C | |
| REXO1 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EloA BP1, ELOABP1, Elongin A binding protein 1, KIAA1138, REX1, REXO1, RNA exonuclease 1 homolog, TCEB3BP1 | |
| Rexo1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title