missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24945SS
This item is not returnable.
View return policy
Description
RGS5 Polyclonal specifically detects RGS5 in Human, Rat, Porcine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RGS5 | |
| Polyclonal | |
| Western Blot 1:250-1: 1000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| O15539 | |
| RGS5 | |
| Within the range of amino acids 169-181 of human RGS5 were used as immunogen. | |
| 0.025 mL | |
| Signal Transduction | |
| 8490 | |
| Human, Rat, Pig, Canine, Primate | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody is reactive to Chimpanzee, Dog, Human, Monkey, Mouse, Pig, Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction