missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 361.00€
Specifications
| Antigen | RGS5 |
|---|---|
| Dilution | Western Blot 1:250-1: 1000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18103092
|
Novus Biologicals
NBP2-24945SS |
0.025 mL |
213.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18766723
|
Novus Biologicals
NBP2-24945 |
0.1 mL |
361.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGS5 Polyclonal specifically detects RGS5 in Human, Rat, Porcine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RGS5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| RGS5 | |
| IgG | |
| Unpurified | |
| This antibody is reactive to Chimpanzee, Dog, Human, Monkey, Mouse, Pig, Rat. |
| Western Blot 1:250-1: 1000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Rat, Pig, Canine, Primate | |
| O15539 | |
| 8490 | |
| Within the range of amino acids 169-181 of human RGS5 were used as immunogen. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title