missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ribonuclease A Monoclonal antibody specifically detects Ribonuclease A in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | Ribonuclease A |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 02 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | ELISA, Sandwich ELISA Capture |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ribonuclease A (Uniprot#: P07998; Met1-Thr156). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?