missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ritanserin

Product Code. 18024192
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024192 50 mg 50mg
18065162 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024192 Supplier Tocris Bioscience™ Supplier No. 1955/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent 5-HT2 antagonist

Potent and long-acting 5-HT2 receptor antagonist (Ki = 0.39 nM). Anxiolytic in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87051-43-2
Molecular Formula C27H25F2N3OS

Specifications

Quantity 50 mg
Formula Weight 477.57
Purity >99%
Molecular Weight (g/mol) 477.574
Chemical Name or Material Ritanserin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.