missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROCK2 Polyclonal Antibody

Description
Positive Control: 293T, A431, HeLa, HepG2, Rat2, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, A549, H1299, HCT116 Predicted Reactivity: Xenopus laevis (80%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ROCK2, the predominant ROCK isoform in skeletal muscle, is an important regulator downstream of Rho GTPases. ROCK2 regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho.
Specifications
Specifications
| Antigen | ROCK2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Gene | ROCK2 |
| Gene Accession No. | O75116, P70336, Q62868 |
| Gene Alias | B230113H15Rik; KIAA0619; mKIAA0619; p150 ROK-alpha; p164 ROCK-2; Rho associated coiled-coil containing protein kinase 2; Rho kinase 2; RhoA - binding serine/threosine kinase alpha (ROK - alpha); rhoA-binding kinase 2; Rho-associated coiled-coil containing protein kinase 2; Rho-associated coiled-coil forming kinage 2; Rho-associated coiled-coil forming kinase 2; rho-associated protein kinase 2; Rho-associated, coiled-coil-containing protein kinase 2; rho-associated, coiled-coil-containing protein kinase II; Rho-kinase; ROCK II; Rock2; Rock2m; rockii; ROCK-II; ROK; ROKalpha |
| Gene Symbols | ROCK2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?