missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
CD102 (ICAM-2) Monoclonal Antibody Specifically detects CD102 (ICAM-2) in Mouse Samples. It is validated for Flow Cytometry.
The 3C4 (mIC2/4) monoclonal antibody specifically binds to the mouse ICAM-2 (CD102) cell surface glycoprotein, which is a ligand for LFA-1. CD102 is constitutively expressed on endothelial cells, T and B lymphocytes, and alveolar walls, and also on various leukocyte cell lines. It does not appear to be involved in the development of hematopoietic cells. In a model for allergic asthma, endothelial CD102 mediates the transmigration of eosinophils (but not lymphocytes, monocytes, or macrophages) into the airway lumen. The 3C4 (mIC2/4) antibody blocks interactions between ICAM-2 and LFA-1.
Specifica
Specifica
| Antigen | CD102 (ICAM-2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3C4(mIC2/4) |
| Concentration | 0.2 mg/mL |
| Conjugate | RR688 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | ICAM-2 |
| Host Species | Rat |
| Immunogen | Transfected Cell Line |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?