missing translation for 'onlineSavingsMsg'
Learn More

(S)-Remoxipride Hydrochloride, TRC

Product Code. 30424836
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424836 100 mg -
30426983 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424836 Supplier Toronto Research Chemicals Supplier No. TRCR143700100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73220-03-8
Molecular Formula C16H24BrClN2O3
Molecular Weight (g/mol) 407.73
Synonym 3-Bromo-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2,6-dimethoxybenzamide Hydrochloride, (-)-Remoxipride Hydrochloride, Remoxipride Hydrochloride, (-)-FLA 731, A 33547, A 33547 Hydrochloride, FLA 731, (-)-FLA 73, Roxiam
IUPAC Name 3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride
SMILES Cl.CCN1CCC[C@H]1CNC(=O)c2c(OC)ccc(Br)c2OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H23BrN2O3.ClH/c1-4-19-9-5-6-11(19)10-18-16(20)14-13(21-2)8-7-12(17)15(14)22-3;/h7-8,11H,4-6,9-10H2,1-3H3,(H,18,20);1H/t11-;/m0./s1
Formula Weight 406.0659
Chemical Name or Material (S)-Remoxipride Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.