missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S100Z Polyclonal Antibody
GREENER_CHOICE

Product Code. 16351045
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16351045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16351045 Supplier Invitrogen™ Supplier No. PA584092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPTQLEMAMD TMIRIFHRYS GKARKRFKLS Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 83%.

Members of the S100 protein family contain 2 calcium-binding EF-hands and exhibit cell-type specific expression patterns. For additional background information on S100 proteins, see MIM 114085.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100Z
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S100Z
Gene Accession No. Q8WXG8
Gene Alias Gm625; Protein S100-Z; S100 calcium binding protein Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z; S100Z; S100-zeta
Gene Symbols S100Z
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human S100Z. Recombinant protein control fragment (Product #RP-108392).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.