missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17723-1-AP-150UL
This item is not returnable.
View return policy
Description
SMD4B has transcriptional repressor activity. Overexpression of SMD4B inhibits the transcriptional activities of AP-1, p53/TP53 and CDKN1A.Specifications
| SAMD4B | |
| Polyclonal | |
| Unconjugated | |
| SAMD4B | |
| hSmaug2, Protein Smaug homolog 2, SAMD4B, Smaug 2, SMAUG2, SMGB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233033, 308473, 55095 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PRF9, Q80XS6 | |
| SAMD4B | |
| SAMD4B Fusion Protein Ag12113 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction