missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SAMD4B |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817674
|
Proteintech
17723-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807674
|
Proteintech
17723-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMD4B has transcriptional repressor activity. Overexpression of SMD4B inhibits the transcriptional activities of AP-1, p53/TP53 and CDKN1A.Specifications
| SAMD4B | |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q5PRF9, Q80XS6 | |
| 233033, 308473, 55095 | |
| SAMD4B Fusion Protein Ag12113 | |
| Primary | |
| -20°C | |
| SAMD4B |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hSmaug2, Protein Smaug homolog 2, SAMD4B, Smaug 2, SMAUG2, SMGB | |
| SAMD4B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title