missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sarcalumenin Monoclonal antibody specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Sarcalumenin |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | XIIC4 |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | sarcalumenin |
| Host Species | Mouse |
| Immunogen | Purified rabbit sarcoplasmic reticulum membrane vesicles. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?