missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11130-100
467.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for ELISA
Specifications
| SARS-CoV-2 Spike S2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 43740568 | |
| SARS-CoV-2 | |
| Purified |
| ELISA | |
| 2812B | |
| ELISA | |
| COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
| Recombinant SARS-CoV2 Spike S2 Subunit, | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction