missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™

Rabbit Monoclonal Antibody
221.00€ - 580.00€
Specifications
| Antigen | SARS-CoV-2 Spike S2 |
|---|---|
| Clone | 2812B |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730917
![]() |
R&D Systems
MAB11130-SP |
25 μg |
221.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761007
|
R&D Systems
MAB11130-100 |
100 μg |
580.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for ELISASpecifications
| SARS-CoV-2 Spike S2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 43740568 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2812B | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| SARS-CoV-2 | |
| COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
| Recombinant SARS-CoV2 Spike S2 Subunit, | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title