missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCML1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13454707
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454707 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454707 Supplier Invitrogen™ Supplier No. PA530165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Rhesus Monkey (89%), Chimpanzee (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. May be involved in spermatogenesis during sexual maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCML1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Scml1
Gene Accession No. Q9UN30
Gene Alias Scm polycomb group protein like 1; SCML1; sex comb on midleg like 1; sex comb on midleg-like 1 (Drosophila); sex comb on midleg-like protein 1
Gene Symbols Scml1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SCML1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.