missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCRT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15778781 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15778781 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15778781 Supplier Invitrogen™ Supplier No. PA560966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LHDKGYLSDY VGPSSVYDGD AEAALLKGPS PEPMYAAAVR GELGP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene is a member of the Snail family of C2H2-type zinc finger transcription factors. It codes for a neural-specific transcriptional repressor that binds to E-box motifs. The protein may promote neural differention and may be involved in cancers with neuroendocrine features.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scrt1
Gene Accession No. Q99M85, Q9BWW7
Gene Alias mScrt; scratch 1; scratch family transcriptional repressor 1; scratch family zinc finger 1; scratch homolog 1 zinc finger protein; scratch homolog 1, zinc finger protein; scratch homolog 1, zinc finger protein (Drosophila); SCRT; Scrt1; Transcriptional repressor scratch 1; ZNF898
Gene Symbols Scrt1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCRT1. Recombinant protein control fragment (Product #RP-100027).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170729, 83482
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.