missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEMA3B Polyclonal Antibody
GREENER_CHOICE

Product Code. 17182184 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17182184 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17182184 Supplier Invitrogen™ Supplier No. PA5114387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human U2OS whole cell, human A549 whole cell, human U-87MG whole cell, rat thymus tissues, rat kidney tissue, mouse thymus tissue, mouse lung tissue, mouse small intestine tissue, mouse kidney tissue, mouse HEPA1-6 whole cel. IHC: human mammary cancer tissue, human colon cancer tissue, mouse brain tissue, mouse brain tissue, rat brain tissue, rat brain tissue. ICC/IF: A431 cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene SEMA3B
Gene Accession No. Q13214, Q62177
Gene Alias LUCA-1; Seam V antibody; SemA; sema A; sema A(V); sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B; Sema V; SEMA3B; SEMA5; SEMAA; semaphorin 3B; semaphorin A; semaphorin-3B; semaphorin-A; semaphorin-V; semaV
Gene Symbols SEMA3B
Host Species Rabbit
Immunogen E.coli-derived human Semaphorin 3B/SEMA3B recombinant protein (Position: Y55-R624).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20347, 363142, 7869
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.