missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERGEF Monoclonal Antibody (1A2)

Description
Peptide Sequence: KHGQLANEAA FLPVPQKIEA HCFQNEKVTA IWSGWTHLVA QTETGKMFTW GRADYGQLGR KLETYEGWKL EKQDSFLPCS RPPNSMPSSP HCLTGATE.
Probable guanine nucleotide exchange factor (GEF), which may be involved in the secretion process.
Specifications
Specifications
| Antigen | SERGEF |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1A2 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SERGEF |
| Gene Accession No. | Q9UGK8 |
| Gene Alias | deafness locus associated putative guanine nucleotide exchange factor; deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Gef; Gnef; Gnefr; guanine nucleotide exchange factor (RCC1 related); guanine nucleotide exchange factor-related protein; secretion regulating guanine nucleotide exchange factor; secretion-regulating guanine nucleotide exchange factor; Sergef |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?