missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SERPINA6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15915545
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915545 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915545 Supplier Invitrogen™ Supplier No. PA579977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HL-60 whole cell, human A375 whole cell, human 293T whole cell. IHC: human liver cancer tissue. ICC/IF: HepG2 cell.

This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SERPINA6
Gene Accession No. P08185
Gene Alias AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin
Gene Symbols SERPINA6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human Cortisol Binding Globulin (102-120aa ETEIHQGFQHLHQLFAKSD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.