missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SFRS15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15533951
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15533951 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15533951 Supplier Invitrogen™ Supplier No. PA536611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 126 kDa. Purity is >95% by SDS-PAGE.

The family of SR factors all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. They are not only essential for constitutive splicing, but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Splicing factor arginine/serine-rich 15 (SFRS15), also designated CTD-binding SR-like protein RA4, contains one RRM and one SR-rich domains. SFRS15 interacts with C-terminal repetitive domain (CTD) of Pol II and is believed to functionally and physically link transcription and pre-mRNA processing. Localized to the nucleus, SFRS15 is expressed as two isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SCAF4
Gene Accession No. O95104, Q63627
Gene Alias AA517739; CTD-binding SR-like protein RA4; KIAA1172; pre-mRNA splicing SR protein rA4; SCAF4; serine/arginine-rich splicing factor 15; SFR15; SFRS15; splicing factor serine alanine 15; splicing factor, arginine/serine-rich 15; SRA4; SR-like CTD-associated factor 4; SR-related and CTD-associated factor 4; SR-related CTD associated factor 4; SR-related CTD-associated factor 4; Srsf15
Gene Symbols SCAF4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 611-660 of human SFRS15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224432, 245924, 57466
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.