Learn More
SH3BP5 Mouse, anti-Human, Unconjugated, Clone: 1B9D4, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SH3BP5 |
|---|---|
| Clone | 1B9D4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17211506
|
Proteintech
67841-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17201506
|
Proteintech
67841-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SH3BP5(SH3 domain-binding protein 5) is also named as SAB. It is previously isolated in a screen for proteins capable of binding to the Src homology 3 (SH3) domain of Bruton's tyrosine kinase (Btk) and could play a role in targeting JNK and Btk to mitochondria and/or mediating cross-talk between the JNK and Btk signalling pathways. It has 2 isoforms produced by alternative splicing with the molecular mass of 50 kDa and 32 kDa. There are two bands of 83 kDa and 70 kDa can be detected in A431 and the larger species may correspond to splice variants of SH3BP5 or SH3BP5 proteins that have undergone post-translational modification(PMID:12167088). In addition, we detect the band of 32 kDa as the isoform 2 of SH3BP5 and another band of 62 kDa besides the 47 kDa and 80 kDa.
Specifications
| SH3BP5 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat, Pig, Chicken | |
| Primary |
| 1B9D4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 9467 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.