missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin, Sheep, Polyclonal Antibody, Abnova™
Specifications
Specifications
| Antigen | Fibronectin |
| Applications | ELISA, Immunocytochemistry, Immunodiffusion, Immunofluorescence, Immunomicroscopy, Radioimmune Assays (RIA) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Sheep polyclonal antibody raised against native Fibronectin. |
| Dilution | ELISA (1:5000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user. |
| Formulation | In serum |
| Host Species | Sheep |
| Immunogen | Native purified from human plasma Fibronectin. |
| Quantity | 1 mL |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?