missing translation for 'onlineSavingsMsg'
Learn More

SR-alpha, Sheep, Polyclonal Antibody, Abnova™

Product Code. 16049246
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16049246 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16049246 Supplier Abnova Supplier No. PAB10405.250ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against partial recombinant SR-alpha.

Specifications

Antigen SR-alpha
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant SR-alpha.
Dilution Western blot (1 to 5 mg/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 39-295 of SR-alpha.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.