Learn More
Silicon powder, crystalline, -325 mesh, 99.5% (metals basis)
Product Code. 11387507
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387507 100 g 100g
11397507 10 g 10g
11377507 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11387507 Supplier Thermo Scientific Alfa Aesar Supplier No. 012681.22
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 7440-21-3 | Si | 28.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]
Specifications
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99.5% (Metals basis)
Odor Odorless
Appearance Powder
Density 2.33g/cm3
Chemical Name or Material Silicon