missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SKA2 Polyclonal specifically detects SKA2 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | SKA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 1 μg/mL, ELISA 1:100-1:2000 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | NP_872426 |
| Gene Alias | FAM33A, family with sequence similarity 33, member A, FLJ12758, MGC110975, spindle and kinetochore associated complex subunit 2, spindle and kinetochore-associated protein 2, spindle and KT (kinetochore) associated 2 |
| Gene Symbols | SKA2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?