Learn More
SLC22A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24617-1-AP-150UL
 View more versions of this product
Product Code. 16893105
  • 410.00€ / 150µL
Description
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
Specifications
Specifications
SLC22A1
Polyclonal
Unconjugated
SLC22A1
HOCT1, OCT1, oct1_cds, Organic cation transporter 1, SLC22A1
Rabbit
Antigen Affinity Chromatography
RUO
20517, 24904, 6580
-20°C
Liquid
Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08966, O15245, Q63089
Slc22a1
SLC22A1 Fusion Protein Ag18806
150 μL
Primary
Human, Mouse, Rat