missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC24A4 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RQRLINSANG VSSKPLQNGR HENIENGNVP VENPEDPQQN QEQQP Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.
This gene encodes a member of the potassium-dependent sodium/calcium exchanger protein family. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | SLC24A4 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC24A4 |
| Gene Accession No. | Q8NFF2 |
| Gene Alias | A930002M03Rik; AI2A5; AW492432; K-dependent Na/Ca exchanger NCKX4; Na(+)/K(+)/Ca(2+)-exchange protein 4; NCKX4; SHEP6; SLC24A2; Slc24a4; Sodium/potassium/calcium exchanger 4; solute carrier family 24 (sodium/potassium/calcium exchanger), member 4; solute carrier family 24 member 4 |
| Gene Symbols | SLC24A4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?